Allylchlorodimethylsilane ≥97%
Supplier: BeanTown Chemical
Synonyms:
Allyldimethylchlorosilane, ADMCS
223775-25G
223775-5G
BT223775-25GEA
230.61
USD
BT223775-25G
BT223775-5G
Allylchlorodimethylsilane ≥97%
Allylchlorodimethylsilane
CAS: 4028-23-3; EC No: 223-706-2; MDL No: MFCD00000506
UN No: UN2985; Haz Class: 3 (8); Packing Group: II
Liquid; Linear Formula: CH2=CHCH2Si(CH3)2Cl; Molecular Formula: C5H11ClSi; MW: 134.68
Boiling Point: 111-113°; Flash point: 5°C (41°F)
Density (g/mL): 0.891; Refractive Index: 1.427
Moisture Sensitive
- Refractive index: 1.427
- Moisture Sensitive
- Liquid
Formula:
C₅H₁₁ClSi MW: 134.68 g/mol Boiling Pt: 111…113 °C Density: 0.891 g/mL Flash Pt: 5 °C (41 °F) |
MDL Number:
MFCD00000506 CAS Number: 4028-23-3 EINECS: 223-706-2 UN: 2985 ADR: 3,II |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...