Zinc tetrafluoroborate hydrate

Supplier: BeanTown Chemical

223755-10G 223755-250G 223755-50G
BT223755-10GEA 23.23 USD
BT223755-10G BT223755-250G BT223755-50G
Zinc tetrafluoroborate hydrate
Zinc tetrafluoroborate hydrate

CAS: 27860-83-9; EC No: 237-534-0; MDL No: MFCD00150371 UN No: UN3260; Haz Class: 8; Packing Group: III Crystalline/Powder; Linear Formula: Zn(BF4)2·xH2O; MW: 238.90 (anhydrous) Hygroscopic


  • Hygroscopic
  • Crystalline/Powder

Formula: Zn(BF₄)₂·xH₂O
MW: 238.98 g/mol (anhy)
MDL Number: MFCD00150371
CAS Number: 27860-83-9
EINECS: 237-534-0
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR