N,N-Dimethylcarbamoyl chloride ≥96%
Supplier: BeanTown Chemical
Synonyms:
Dimethylcarbamyl chloride
223600-100G
223600-25G
BT223600-100GEA
42.91
USD
BT223600-100G
BT223600-25G
N,N-Dimethylcarbamoyl chloride ≥96%
N,N-Dimethylcarbamoyl chloride
CAS: 79-44-7; EC No: 201-208-6; MDL No: MFCD00000635; RTECS: FD4200000
UN No: UN2262; Haz Class: 8; Packing Group: II
Liquid; Linear Formula: (CH3)2NCOCl; Molecular Formula: C3H6ClNO; MW: 107.54
Melting Point: -33°; Boiling Point: 167-168°/775 mmHg; Flash point: 82°C (180°F)
Density (g/mL): 1.168; Refractive Index: 1.453
Moisture Sensitive
- RTECS: FD4200000
- Refractive index: 1.453
- Moisture Sensitive
- Liquid
Formula:
C₃H₆ClNO MW: 107.54 g/mol Boiling Pt: 167…168 °C/775 mmHg Melting Pt: –33 °C Density: 1.168 g/mL Flash Pt: 82 °C (180 °F) |
MDL Number:
MFCD00000635 CAS Number: 79-44-7 EINECS: 201-208-6 UN: 2262 ADR: 8,II |
Specification Test Results
Assay | 96% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...