Hydroxylamine-O-sulphonic acid ≥97%
Supplier: BeanTown Chemical
Synonyms:
Amino hydrogen sulphate
223535-100G
223535-25G
BT223535-100GEA
231
USD
BT223535-100G
BT223535-25G
Hydroxylamine-O-sulphonic acid ≥97%
Hydroxylamine-O-sulphonic acid
CAS: 2950-43-8; EC No: 220-971-6; MDL No: MFCD00011604; RTECS: NC5427000
UN No: UN3260; Haz Class: 8; Packing Group: III
Solid; Linear Formula: NH2OSO3H; MW: 113.09
Melting Point: 210° (decomposes)
Hygroscopic
- RTECS: NC5427000
- Hygroscopic
- Solid
Formula:
H₃NO₄S Melting Pt: 210 °C (dec.) |
MDL Number:
MFCD00011604 CAS Number: 2950-43-8 EINECS: 220-971-6 UN: 3260 ADR: 8,III |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...