Potassium hexafluorozirconate ≥99%
Supplier: BeanTown Chemical
223525-2KG
223525-500G
BT223525-2KGEA
272.52
USD
BT223525-2KG
BT223525-500G
Potassium hexafluorozirconate ≥99%
Potassium hexafluorozirconate
CAS: 16923-95-8; EC No: 240-985-6; MDL No: MFCD00042547
UN No: UN3288; Haz Class: 6.1; Packing Group: III
-10 Mesh Powder; Linear Formula: K2ZrF6; MW: 283.41
Density (g/mL): 3.48
Hygroscopic
- Hygroscopic
- -10 Mesh Powder
Formula:
F₆K₂Zr MW: 283.41 g/mol Density: 3.48 g/mL |
MDL Number:
MFCD00042547 CAS Number: 16923-95-8 EINECS: 240-985-6 UN: 3288 ADR: 6.1,III |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...