Diethyl fluoromalonate ≥97%

Supplier: BeanTown Chemical

Synonyms: 1,3-diethyl 2-fluoropropanedioate

223500-1G 223500-5G
BT223500-1GEA 83 USD
BT223500-1G BT223500-5G
Diethyl fluoromalonate ≥97%
Diethyl fluoromalonate

CAS: 685-88-1; EC No: 211-684-7; MDL No: MFCD00009139; RTECS: OO1670000 UN No: UN1760; Haz Class: 8; Packing Group: II Liquid; Linear Formula: FCH(CO2CH2CH3)2; Molecular Formula: C7H11FO4; MW: 178.16 Boiling Point: 121-122°/30 mmHg; Flash point: 62°C (143°F) Density (g/mL): 1.129; Refractive Index: 1.407


  • RTECS: OO1670000
  • Refractive index: 1.407
  • Liquid

Formula: C₇H₁₁FO₄
MW: 178.16 g/mol
Boiling Pt: 121…122 °C/30 mmHg
Density: 1.129 g/mL
Flash Pt: 62 °C (143 °F)
MDL Number: MFCD00009139
CAS Number: 685-88-1
EINECS: 211-684-7
ADR: 8,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR