2-Cyclohexen-1-one ≥97%
Supplier: BeanTown Chemical
223455-100G
223455-25G
223455-500G
BT223455-100GEA
103
USD
BT223455-100G
BT223455-25G
BT223455-500G
2-Cyclohexen-1-one ≥97%
2-Cyclohexen-1-one
CAS: 930-68-7; EC No: 213-223-5; MDL No: MFCD00001577; RTECS: GW7000000
UN No: UN2929; Haz Class: 6.1 (3); Packing Group: II
Liquid; Molecular Formula: C6H8O; MW: 96.13
Melting Point: -53°; Boiling Point: 171-173°; Flash point: 56°C (133°F)
Density (g/mL): 0.993; Refractive Index: 1.488
- RTECS: GW7000000
- Refractive index: 1.488
- Liquid
Formula:
C₆H₈O MW: 96.13 g/mol Boiling Pt: 171…173 °C Melting Pt: –53 °C Density: 0.993 g/mL Flash Pt: 56 °C (133 °F) |
MDL Number:
MFCD00001577 CAS Number: 930-68-7 EINECS: 213-223-5 UN: 2929 ADR: 6.1,II |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...