2-Methylpiperidine ≥99%
Supplier: BeanTown Chemical
223445-100ML
223445-500ML
BT223445-100MLEA
37
USD
BT223445-100ML
BT223445-500ML
2-Methylpiperidine ≥99%
2-Methylpiperidine
CAS: 109-05-7; EC No: 203-642-1; MDL No: MFCD00005982; RTECS: TN1240000
UN No: UN2733; Haz Class: 3 (8); Packing Group: II
Liquid; Molecular Formula: C6H13N; MW: 99.17
Boiling Point: 118-119°/753 mmHg; Flash point: 10°C (50°F)
Density (g/mL): 0.844; Refractive Index: 1.445
- RTECS: TN1240000
- Refractive index: 1.445
- Liquid
Formula:
C₆H₁₃N MW: 99.18 g/mol Boiling Pt: 118…119 °C/753 mmHg Density: 0.844 g/mL Flash Pt: 10 °C (50 °F) |
MDL Number:
MFCD00005982 CAS Number: 109-05-7 EINECS: 203-642-1 ADR: 3 (8),II |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...