4-Penten-1-ol ≥98%
Supplier: BeanTown Chemical
Synonyms:
Pent-4-en-1-ol, 1-Penten-5-ol, 4-Penten-1-ol
223440-100G
223440-25G
223440-5G
BT223440-100GEA
545.62
USD
BT223440-100G
BT223440-25G
BT223440-5G
4-Penten-1-ol ≥98%
4-Penten-1-ol
CAS: 821-09-0; EC No: 212-473-2; MDL No: MFCD00002975
UN No: UN1987; Haz Class: 3; Packing Group: III
Liquid; Linear Formula: H2C=CH(CH2)3OH; Molecular Formula: C5H10O; MW: 86.13
Boiling Point: 134-137°; Flash point: 48°C (118°F)
Density (g/mL): 0.834; Refractive Index: 1.429
- Refractive index: 1.429
- Liquid
Formula:
C₅H₁₀O MW: 86.13 g/mol Boiling Pt: 134…137 °C Density: 0.834 g/mL Flash Pt: 48 °C (118 °F) |
MDL Number:
MFCD00002975 CAS Number: 821-09-0 EINECS: 212-473-2 UN: 1987 ADR: 3,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...