Indium(III) chloride hydrate ≥99.99%
Supplier: BeanTown Chemical
223410-10G
223410-250G
223410-50G
BT223410-10GEA
76
USD
BT223410-10G
BT223410-250G
BT223410-50G
Indium(III) chloride hydrate ≥99.99%
Indium(III) chloride hydrate
CAS: 143983-91-9; EC No: 233-043-0; MDL No: MFCD00011058
UN No: UN3260; Haz Class: 8; Packing Group: II
Crystalline; Linear Formula: InCl3·xH2O; MW: 221.18 (anhydrous)
Hygroscopic
- Hygroscopic
- Crystalline
Formula:
Cl₃In·H₂O MW: 221.18 g/mol (anhy) |
MDL Number:
MFCD00011058 CAS Number: 143983-91-9 EINECS: 233-043-0 UN: 3260 ADR: 8 II Merck Index: 13,04980 |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...