3-Butyne-1-ol ≥98%
Supplier: BeanTown Chemical
Synonyms:
4-Hydroxy-1-butyne, But-3-yn-1-ol, 3-Butyn-1-ol, 1-Hydroxy-3-butyne
223320-250G
223320-25G
223320-5G
BT223320-250GEA
1112
USD
BT223320-250G
BT223320-25G
BT223320-5G
3-Butyne-1-ol ≥98%
3-Butyne-1-ol
CAS: 927-74-2; EC No: 213-161-9; MDL No: MFCD00002955; RTECS: ES0710000
UN No: UN1987; Haz Class: 3; Packing Group: III
Liquid; Linear Formula: HC≡CCH2CH2OH; Molecular Formula: C4H6O; MW: 70.09
Melting Point: -64°; Boiling Point: 129°; Flash point: 36°C (97°F)
Density (g/mL): 0.926; Refractive Index: 1.441
- RTECS: ES0710000
- Refractive index: 1.441
- Liquid
Formula:
C₄H₆O MW: 70.09 g/mol Boiling Pt: 129 °C Melting Pt: –64 °C Density: 0.926 g/mL Flash Pt: 36 °C (97 °F) |
MDL Number:
MFCD00002955 CAS Number: 927-74-2 EINECS: 213-161-9 UN: 1987 ADR: 3,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...