(±)-1-Ethoxy-2-propanol ≥90% 2-ethoxy-1-propanol main impurity, Technical Grade

Supplier: BeanTown Chemical

223310-4X500ML 223310-500ML
BT223310-4X500EA 153 USD
BT223310-4X500 BT223310-500ML
(±)-1-Ethoxy-2-propanol ≥90% 2-ethoxy-1-propanol main impurity, Technical Grade
(±)-1-Ethoxy-2-propanol

CAS: 1569-02-4; EC No: 216-374-5; MDL No: MFCD00067050 UN No: UN1987; Haz Class: 3; Packing Group: III Liquid; Molecular Formula: C5H12O2; MW: 104.15 Flash point: 42°C (108°F) Density (g/mL): 0.896


  • Liquid

Formula: C₅H₁₂O₂
Density: 0.896 g/mL
Flash Pt: 42 °C (108 °F)
MDL Number: MFCD00067050
CAS Number: 1569-02-4
EINECS: 216-374-5
UN: 1987
ADR: 3,III

Order Now

Specification Test Results

Assay 90%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR