Cyclopropanecarboxylic acid ≥98%

Supplier: BeanTown Chemical

223260-1KG 223260-250G 223260-50G
BT223260-1KGEA 414 USD
BT223260-1KG BT223260-250G BT223260-50G
Cyclopropanecarboxylic acid ≥98%
Cyclopropanecarboxylic acid

CAS: 1759-53-1; EC No: 217-162-5; MDL No: MFCD00001287; RTECS: GZ1100000 UN No: UN3265 ; Haz Class: 8; Packing Group: II Liquid; Molecular Formula: C4H6O2 ; MW: 86.09 Melting Point: 14-17°; Boiling Point: 182-184°; Flash point: 72°C (162°F) Density (g/mL): 1.081; Refractive Index: 1.438


  • RTECS: GZ1100000
  • Refractive index: 1.438
  • Liquid

Formula: C₄H₆O₂
MW: 86.09 g/mol
Boiling Pt: 182…184 °C
Melting Pt: 14…17 °C
Density: 1.081 g/mL
Flash Pt: 72 °C (162 °F)
MDL Number: MFCD00001287
CAS Number: 1759-53-1
EINECS: 217-162-5
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR