Triethoxy-1H,1H,2H,2H-heptadecafluorodecylsilane ≥97%
Supplier: BeanTown Chemical
Synonyms:
1H,1H,2H,2H-Perfluorodecyltriethoxysilane, Triethoxy(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl)silane, Triethoxy-1H,1H,2H,2H-perfluorodecylsilane
223050-1G
223050-25G
223050-5G
BT223050-1GEA
36
USD
BT223050-1G
BT223050-25G
BT223050-5G
Triethoxy-1H,1H,2H,2H-heptadecafluorodecylsilane ≥97%
Triethoxy-1H,1H,2H,2H-heptadecafluorodecylsilane
CAS: 101947-16-4; MDL No: MFCD00042334
UN No: UN1760; Haz Class: 8; Packing Group: III
Liquid; Molecular Formula: C16F17H19O3Si; MW: 610.38
Boiling Point: 209-230°; Flash point: 110°C (230°F)
Density (g/mL): 1.389; Refractive Index: 1.342
Moisture Sensitive
- Refractive index: 1.342
- Moisture Sensitive
- Liquid
Formula:
C₁₆H₁₉F₁₇O₃Si Boiling Pt: 209…230 °C Density: 1.389 g/mL Flash Pt: 110 °C (230 °F) |
MDL Number:
MFCD00042334 CAS Number: 101947-16-4 ADR: 8,III |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...