3-Aminopyridine ≥99%

Supplier: BeanTown Chemical

Synonyms: Pyridin-3-amine, 3-Pyridylamine

223040-1KG 223040-250G 223040-50G
BT223040-1KGEA 340 USD
BT223040-1KG BT223040-250G BT223040-50G
3-Aminopyridine ≥99%
3-Aminopyridine

CAS: 462-08-8; EC No: 207-322-2; MDL No: MFCD00006400; RTECS: US1650000 UN No: UN2671; Haz Class: 6.1; Packing Group: II Flakes; Molecular Formula: C5H6N2 ; MW: 94.11 Melting Point: 60-63°; Boiling Point: 248°; Flash point: 88°C (190°F) Air Sensitive , Hygroscopic


  • RTECS: US1650000
  • Air Sensitive, Hygroscopic
  • Flakes

Formula: C₅H₆N₂
MW: 94.12 g/mol
Boiling Pt: 248 °C
Melting Pt: 60…63 °C
Flash Pt: 88 °C (190 °F)
MDL Number: MFCD00006400
CAS Number: 462-08-8
EINECS: 207-322-2
UN: 2671
ADR: 6.1,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR