Sodium hexafluorophosphate ≥99%
Supplier: BeanTown Chemical
223025-100G
223025-25G
BT223025-100GEA
352
USD
BT223025-100G
BT223025-25G
Sodium hexafluorophosphate ≥99%
Sodium hexafluorophosphate
CAS: 21324-39-0; EC No: 244-333-1; MDL No: MFCD00011122
UN No: UN3260; Haz Class: 8; Packing Group: II
Powder; Linear Formula: NaPF6; MW: 167.95
Melting Point:
- Hygroscopic
- Powder
Formula:
F₆NaP Melting Pt: >200 °C Density: 2.369 g/mL |
MDL Number:
MFCD00011122 CAS Number: 21324-39-0 EINECS: 244-333-1 UN: 3260 ADR: 8,II |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...