1-(Trimethylsilyl)-1-propyne ≥98%

Supplier: BeanTown Chemical

Synonyms: Trimethyl-1-propynylsilane

223000-1G 223000-25G 223000-5G
BT223000-1GEA 48 USD
BT223000-1G BT223000-25G BT223000-5G
1-(Trimethylsilyl)-1-propyne ≥98%
1-(Trimethylsilyl)-1-propyne

CAS: 6224-91-5; EC No: 228-314-5; MDL No: MFCD00009271 UN No: UN1993; Haz Class: 3; Packing Group: II Liquid; Linear Formula: CH3C≡CSi(CH3)3; Molecular Formula: C6H12Si; MW: 112.24 Boiling Point: 99-100°; Flash point: 4°C (39°F) Density (g/mL): 0.758; Refractive Index: 1.417


  • Refractive index: 1.417
  • Liquid

Formula: C₆H₁₂Si
MW: 112.25 g/mol
Boiling Pt: 99…100 °C
Density: 0.758 g/mL
Flash Pt: 4 °C (39 °F)
MDL Number: MFCD00009271
CAS Number: 6224-91-5
EINECS: 228-314-5
ADR: 3,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR