1-(4-Fluorophenyl)-2-methyl-2-propylamine ≥96%

Supplier: BeanTown Chemical

222975-1G 222975-5G
BT222975-1GEA 92 USD
BT222975-1G BT222975-5G
1-(4-Fluorophenyl)-2-methyl-2-propylamine ≥96%
1-(4-Fluorophenyl)-2-methyl-2-propylamine

CAS: 1200-27-7; EC No: 000-000-0; MDL No: MFCD00082702 UN No: UN2735; Haz Class: 8; Packing Group: III Liquid; Molecular Formula: C10H14FN; MW: 167.23 Flash point: 110°C (230°F)


  • Liquid

Formula: C₁₀H₁₄FN
Flash Pt: 110 °C (230 °F)
MDL Number: MFCD00082702
CAS Number: 1200-27-7
ADR: 8,III

Order Now

Specification Test Results

Assay 96%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR