1-Bromo-3,4,5-trifluorobenzene ≥98%
Supplier: BeanTown Chemical
Synonyms:
5-Bromo-1,2,3-trifluorobenzene
222945-1G
222945-5G
BT222945-1GEA
52
USD
BT222945-1G
BT222945-5G
1-Bromo-3,4,5-trifluorobenzene ≥98%
1-Bromo-3,4,5-trifluorobenzene
CAS: 138526-69-9; EC No: 418-480-9; MDL No: MFCD00070738
UN No: UN1993 ; Haz Class: 3; Packing Group: III
Liquid; Molecular Formula: C6H2BrF3; MW: 210.98
Boiling Point: 47-49°/60 mmHg; Flash point: 45°C (113°F)
Density (g/mL): 1.767; Refractive Index: 1.482
- Refractive index: 1.482
- Liquid
Formula:
C₆H₂BrF₃ MW: 210.98 g/mol Boiling Pt: 47…49 °C/60 mmHg Density: 1.767 g/mL Flash Pt: 45 °C (113 °F) |
MDL Number:
MFCD00070738 CAS Number: 138526-69-9 EINECS: 418-480-9 UN: 1993 ADR: 3,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...