Potassium bromate ≥99.8% ACS
Supplier: BeanTown Chemical
222900-100G
222900-500G
BT222900-100GEA
66.32
USD
BT222900-100G
BT222900-500G
Potassium bromate ≥99.8% ACS
Potassium bromate
CAS: 7758-01-2; EC No: 231-829-8; MDL No: MFCD00011359; RTECS: EF8725000
UN No: UN1484; Haz Class: 5.1; Packing Group: II
Granular; Linear Formula: KBrO3; MW: 167.00
Melting Point: 350°
Density (g/mL): 3.27
- RTECS: EF8725000
- Granular
Formula:
KBrO₃ Melting Pt: 350 °C Density: 3.27 g/mL |
MDL Number:
MFCD00011359 CAS Number: 7758-01-2 EINECS: 231-829-8 UN: 1484 ADR: 5.1,II |
Specification Test Results
Assay | 99.8% min. |
pH of 5% solution at 25°C | 5.0-9.0 |
Insoluble matter | ≤0.0005% |
Bromide (Br) | Passes test |
Sulfate (SO4) | ≤0.005% |
Heavy metals (as Pb) | ≤0.0005% |
Iron (Fe) | ≤0.002% |
Sodium (Na) | ≤0.01% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...