Iron(II) oxalate hydrate ≥96%
Supplier: BeanTown Chemical
|
Warning
|
Synonyms:
Oxalic acid iron (II) salt, Ferrous oxalate, Iron (II) ethanedioate
222890-2KG
222890-500G
BT222890-2KGEA
170
USD
BT222890-2KG
BT222890-500G
Iron(II) oxalate hydrate ≥96%
Iron(II) oxalate hydrate
CAS: 516-03-0; EC No: 208-217-4; MDL No: MFCD00012475
UN No: UN3288; Haz Class: 6.1; Packing Group: III
Powder; Linear Formula: FeC2O4·xH2O; MW: 143.87 (anhydrous)
Density (g/mL): 2.28
Hygroscopic
- Powder
- Hygroscopic
Formula:
FeC₂O₄·xH₂O MW: 143.87 Density: 2.28 g/mL |
MDL Number:
MFCD00012475 CAS Number: 516-03-0 UN: 3288 ADR: 6.1 III Merck Index: 12,04099 |
Specification Test Results
Assay | 96% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...