Ethyl-2-chloroacetoacetate ≥96%
Supplier: BeanTown Chemical
Synonyms:
Ethyl 2-chloroacetoacetate
222800-100G
222800-500G
BT222800-100GEA
35
USD
BT222800-100G
BT222800-500G
Ethyl-2-chloroacetoacetate ≥96%
Ethyl-2-chloroacetoacetate
CAS: 609-15-4; EC No: 210-180-4; MDL No: MFCD00009141
UN No: UN2920; Haz Class: 8 (3); Packing Group: II
Liquid; Molecular Formula: C6H9ClO3 ; MW: 164.59
Boiling Point: 107°/14 mmHg; Flash point: 54°C (129°F)
Density (g/mL): 1.19; Refractive Index: 1.441
- Refractive index: 1.441
- Liquid
Formula:
C₆H₉ClO₃ MW: 164.59 g/mol Boiling Pt: 107 °C/14 mmHg Density: 1.19 g/mL Flash Pt: 54 °C (129 °F) |
MDL Number:
MFCD00009141 CAS Number: 609-15-4 EINECS: 210-180-4 ADR: 8 (3),II |
Specification Test Results
Assay | 96% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...