Ruthenium(III) chloride hydrate ≥99.99%
Supplier: BeanTown Chemical
Synonyms:
Ruthenium(III) chloride xhydrate, Ruthenium trichloride hydrate
222790-1G
222790-5G
BT222790-1GEA
76.28
USD
BT222790-1G
BT222790-5G
Ruthenium(III) chloride hydrate ≥99.99%
Ruthenium(III) chloride hydrate
CAS: 14898-67-0; EC No: 233-167-5; MDL No: MFCD00149844; RTECS: VM2650000
UN No: UN3260; Haz Class: 8; Packing Group: III
Crystalline; Linear Formula: RuCl3·H2O; MW: 207.43 (anhydrous)
Melting Point: 100° (decomposes)
Hygroscopic
- RTECS: VM2650000
- Hygroscopic
- Crystalline
Formula:
Cl₃Ru·xH₂O MW: 225.44 Melting Pt: 100 °C (dec.) Density: 3.11 |
MDL Number:
MFCD00149844 CAS Number: 14898-67-0 EINECS: 233-167-5 UN: 3260 ADR: 8 III |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...