2,4-Dichloro-5-thiazolecarboxaldehyde ≥96%

Supplier: BeanTown Chemical

Synonyms: 2,4-Dichlorothiazole-5-carbaldehyde

222775-1G 222775-250MG
BT222775-1GEA 335 USD
BT222775-1G BT222775-250MG
2,4-Dichloro-5-thiazolecarboxaldehyde ≥96%
2,4-Dichloro-5-thiazolecarboxaldehyde

CAS: 92972-48-0; MDL No: MFCD00793013 UN No: UN2811; Haz Class: 6.1; Packing Group: III Solid; Molecular Formula: C4HCl2NOS; MW: 182.03 Melting Point: 47-53°


  • Solid

Formula: C₄HCl₂NOS
MW: 182.03 g/mol
Melting Pt: 47…53 °C
MDL Number: MFCD00793013
CAS Number: 92972-48-0
ADR: 6.1,III

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR