Lead(II) telluride ≥99.99%

Supplier: BeanTown Chemical

222770-10G 222770-50G
BT222770-10GEA 125 USD
BT222770-10G BT222770-50G
Lead(II) telluride ≥99.99%
Lead(II) telluride

CAS: 1314-91-6; EC No: 215-247-1; MDL No: MFCD00016274 UN No: UN3284; Haz Class: 6.1; Packing Group: III -325 Mesh Powder; Molecular Formula: PbTe ; MW: 334.79 Melting Point: 905° Density (g/mL): 8.164


  • -325 Mesh Powder

Formula: PbTe
MW: 334.8
Melting Pt: 940
Density: 8.164 g/mL
MDL Number: MFCD00016274
CAS Number: 1314-91-6
EINECS: 215-247-1
UN: 3284
ADR: 6.1 III
Merck Index: 12,05446

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR