Aluminium nitrate nonahydrate ≥98%
Supplier: BeanTown Chemical
222760-1KG
222760-250G
BT222760-1KGEA
124.49
USD
BT222760-1KG
BT222760-250G
Aluminium nitrate nonahydrate ≥98%
Aluminium nitrate nonahydrate
CAS: 7784-27-2; EC No: 236-751-8; MDL No: MFCD00003419; RTECS: BD1050000
UN No: UN1438; Haz Class: 5.1; Packing Group: III
Crystalline; Linear Formula: Al(NO3)3·9H2O ; MW: 375.13
Melting Point: 73°
Hygroscopic
- RTECS: BD1050000
- Hygroscopic
- Crystalline
Formula:
Al(NO₃)₃·9H₂O Melting Pt: 73 °C |
MDL Number:
MFCD00149132 CAS Number: 7784-27-2 EINECS: 236-751-8 UN: 1438 ADR: 5.1,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...