Ammonium sulphide 20 - 24% in water

Supplier: BeanTown Chemical

222720-100ML 222720-500ML
BT222720-100MLEA 84.87 USD
BT222720-100ML BT222720-500ML
Ammonium sulphide 20 - 24% in water
Ammonium sulphide

CAS: 12135-76-1; EC No: 235-223-4; MDL No: MFCD00010892 UN No: UN2683; Haz Class: 8 (6.1,3); Packing Group: II Liquid; Linear Formula: (NH4)2S ; MW: 68.14


  • Liquid

Formula: H₈N₂S
MW: 68.14
Melting Pt: -18
Density: 0.99
MDL Number: MFCD00010892
CAS Number: 12135-76-1
EINECS: 235-223-4
UN: 2683
ADR: 8 II
Merck Index: 13,00560

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR