2-Hepten-4-ol mixture E/Z ≥98%
Supplier: BeanTown Chemical
222685-1G
222685-25G
222685-5G
BT222685-1GEA
31
USD
BT222685-1G
BT222685-25G
BT222685-5G
2-Hepten-4-ol mixture E/Z ≥98%
2-Hepten-4-ol
CAS: 4798-59-8; MDL No: MFCD00021932
UN No: UN1987; Haz Class: 3; Packing Group: III
Liquid; Molecular Formula: C7H14O; MW: 114.19
- Liquid
Formula:
C₇H₁₄O MW: 114.18 g/mol |
MDL Number:
MFCD00021932 CAS Number: 4798-59-8 ADR: 3,III |
Specification Test Results
Assay | 98% |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...