Silver chloride ≥99.995%

Supplier: BeanTown Chemical

222615-100G 222615-25G 222615-5G
BT222615-100GEA 423.67 USD
BT222615-100G BT222615-25G BT222615-5G
Silver chloride ≥99.995%
Silver chloride

CAS: 7783-90-6; EC No: 232-033-3; MDL No: MFCD00003399; RTECS: VW3563000 UN No: UN3077; Haz Class: 9; Packing Group: III Powder; Molecular Formula: AgCl; MW: 143.32 Melting Point: 455°; Boiling Point: 1550° Density (g/mL): 5.56; Refractive Index: 2.067 Light Sensitive, Moisture Sensitive


  • RTECS: VW3563000
  • Refractive index: 2.067
  • Light Sensitive, Moisture Sensitive
  • Powder

Formula: AgCl
MW: 143.32 g/mol
Boiling Pt: 1554
Melting Pt: 455
Density: 5.56 g/mL
MDL Number: MFCD00003399
CAS Number: 7783-90-6
EINECS: 232-033-3
ADR: 9 III
Merck Index: 13,08582

Order Now

Specification Test Results

Assay 99.995%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR