Samarium cobalt
Supplier: BeanTown Chemical
222575-250G
222575-50G
BT222575-250GEA
325
USD
BT222575-250G
BT222575-50G
Samarium cobalt
Samarium cobalt
CAS: 12017-68-4; EC No: 234-625-7; MDL No: MFCD02091682
UN No: UN3089; Haz Class: 4.1; Packing Group: III
Solid, Intermetallic, Magnet Alloy Additive; Linear Formula: SmCo5; MW: 445.03
Air Sensitive, Moisture Sensitive
- Air Sensitive, Moisture Sensitive
- Solid, Intermetallic, Magnet Alloy Additive
Formula:
CoSm MW: 209.293 g/mol |
MDL Number:
MFCD02091682 CAS Number: 12017-68-4 EINECS: 234-625-7 ADR: 4.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...