α,2-Dibromotoluene ≥98%
Supplier: BeanTown Chemical
Synonyms:
2-Bromobenzyl bromide
222555-10G
222555-250G
222555-50G
BT222555-10GEA
48
USD
BT222555-10G
BT222555-250G
BT222555-50G
α,2-Dibromotoluene ≥98%
α,2-Dibromotoluene
CAS: 3433-80-5; EC No: 222-334-8; MDL No: MFCD00000173
UN No: UN1759; Haz Class: 8; Packing Group: II
Solid; Molecular Formula: C7H6Br2; MW: 249.93
Melting Point: 29-32°; Boiling Point: 129°/19 mmHg; Flash point:
- Refractive index: 1.619
- Solid
Formula:
C₇H₆Br₂ MW: 249.93 g/mol Boiling Pt: 129 °C/19 mmHg Melting Pt: 29…32 °C Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00000173 CAS Number: 3433-80-5 EINECS: 222-334-8 UN: 1759 ADR: 3,II |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...