3-Bromoaniline ≥98%
Supplier: BeanTown Chemical
222500-100G
222500-25G
222500-500G
BT222500-100GEA
152
USD
BT222500-100G
BT222500-25G
BT222500-500G
3-Bromoaniline ≥98%
3-Bromoaniline
CAS: 591-19-5; EC No: 209-704-4; MDL No: MFCD00007757; RTECS: CX9855300
UN No: UN2810; Haz Class: 6.1; Packing Group: III
Liquid; Molecular Formula: C6H6BrN; MW: 172.02
Melting Point: 15-18°; Boiling Point: 251°; Flash point: 110°C (230°F)
Density (g/mL): 1.58; Refractive Index: 1.625
- RTECS: CX9855300
- Refractive index: 1.625
- Liquid
Formula:
C₆H₆BrN MW: 172.02 g/mol Boiling Pt: 251 °C Melting Pt: 15…18 °C Density: 1.58 g/mL Flash Pt: 110 °C (230 °F) |
MDL Number:
MFCD00007757 CAS Number: 591-19-5 EINECS: 209-704-4 UN: 2810 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...