4-Aminothiophenol ≥97%

Supplier: BeanTown Chemical

Synonyms: 4-Aminobenzenethiol, 4-Aminophenyl mercaptan, 4-Mercaptoaniline

222495-100G 222495-25G 222495-5G
BT222495-100GEA 544 USD
BT222495-100G BT222495-25G BT222495-5G
4-Aminothiophenol ≥97%
4-Aminothiophenol

CAS: 1193-02-8; EC No: 214-763-4; MDL No: MFCD00007880; RTECS: DC0602500 UN No: UN3263; Haz Class: 8; Packing Group: III Solid; Molecular Formula: C6H7NS; MW: 125.19 Melting Point: 37-42°; Boiling Point: 140-145°/16 mmHg; Flash point: 110°C (230°F) Air Sensitive


  • RTECS: DC0602500
  • Air Sensitive
  • Solid

Formula: C₆H₇NS
MW: 125.19 g/mol
Boiling Pt: 140…145 °C/16 mmHg
Melting Pt: 37…42 °C
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00007880
CAS Number: 1193-02-8
EINECS: 214-763-4
UN: 3263
ADR: 8,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR