2-Bromo-2-methylpropane ≥98% stabilized

Supplier: BeanTown Chemical

222435-100G 222435-500G
BT222435-100GEA 69.56 USD
BT222435-100G BT222435-500G
2-Bromo-2-methylpropane ≥98% stabilized
2-Bromo-2-methylpropane

CAS: 507-19-7; EC No: 208-065-9; MDL No: MFCD00000125; RTECS: TX4150000 UN No: UN2342; Haz Class: 3; Packing Group: II Liquid; Linear Formula: (CH3)3CBr; MW: 137.02 Melting Point: -20°; Boiling Point: 71-73°; Flash point: 16°C (61°F) Density (g/mL): 1.22; Refractive Index: 1.428


  • RTECS: TX4150000
  • Refractive index: 1.428
  • Liquid

Formula: C₄H₉Br
Boiling Pt: 71…73 °C
Melting Pt: –20 °C
Density: 1.22 g/mL
Flash Pt: 16 °C (61 °F)
MDL Number: MFCD00000125
CAS Number: 507-19-7
EINECS: 208-065-9
UN: 2342
ADR: 3,II

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR