Sodium hydrogen sulphate ≥90%, Technical Grade
Supplier: BeanTown Chemical
Synonyms:
Sodium bisulphate
222345-1KG
222345-250G
222345-5KG
BT222345-1KGEA
66.82
USD
BT222345-1KG
BT222345-250G
BT222345-5KG
Sodium hydrogen sulphate ≥90%, Technical Grade
Sodium hydrogen sulphate
CAS: 7681-38-1; EC No: 231-665-7; MDL No: MFCD00003531; RTECS: VZ1860000
UN No: UN3260; Haz Class: 8; Packing Group: III
Crystalline; Linear Formula: NaHSO4; MW: 120.06
Melting Point: 315°
Density (g/mL): 2.435
Hygroscopic
- RTECS: VZ1860000
- Hygroscopic
- Crystalline
Formula:
HNaO₄S MW: 120.06 g/mol Melting Pt: 181.85 Density: 2.435 g/mL |
MDL Number:
MFCD00003531 CAS Number: 7681-38-1 EINECS: 231-665-7 UN: 3260 ADR: 8 III Merck Index: 12,08729 |
Specification Test Results
Assay | 90% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...