Iridium(IV) oxide ≥99.99% (trace metals basis)
Supplier: BeanTown Chemical
222340-1G
222340-5G
BT222340-1GEA
414
USD
BT222340-1G
BT222340-5G
Iridium(IV) oxide ≥99.99% (trace metals basis)
Iridium(IV) oxide
CAS: 12030-49-8; EC No: 234-743-9; MDL No: MFCD00011065
UN No: UN1479; Haz Class: 5.1; Packing Group: II
Powder; Molecular Formula: IrO2; MW: 224.21
- Powder
Formula:
IrO₂ |
MDL Number:
MFCD00011065 CAS Number: 12030-49-8 EINECS: 234-743-9 UN: 1479 ADR: 5.1,II |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...