Methanol-[D₄] ≥99.8% (Isotopic)

Supplier: BeanTown Chemical

222300-1G 222300-25G 222300-5G
BT222300-1GEA 26.11 USD
BT222300-1G BT222300-25G BT222300-5G
Methanol-[D₄] ≥99.8% (Isotopic)
Methanol-[D₄]

CAS: 811-98-3; EC No: 212-378-6; MDL No: MFCD00044637 UN No: UN1230; Haz Class: 3; Packing Group: II Liquid; Linear Formula: CD3OD; MW: 36.07 Melting Point: -98°; Boiling Point: 65.4°; Flash point: 11°C (54°F) Density (g/mL): 0.888; Refractive Index: 1.326 Hygroscopic


  • Refractive index: 1.326
  • Hygroscopic
  • Liquid

Formula: D₃COD
Boiling Pt: 65.4 °C
Melting Pt: –98 °C
Density: 0.888 g/mL
Flash Pt: 11 °C (54 °F)
MDL Number: MFCD00044637
CAS Number: 811-98-3
EINECS: 212-378-6
UN: 1230
ADR: 3,II

Order Now

Specification Test Results

Assay 99.8%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR