Ethyl malonyl chloride ≥95%
Supplier: BeanTown Chemical
Synonyms:
Ethyl Malonyl Chloride
222190-100G
222190-25G
222190-5G
BT222190-100GEA
639
USD
BT222190-100G
BT222190-25G
BT222190-5G
Ethyl malonyl chloride ≥95%
Ethyl malonyl chloride
CAS: 36239-09-5; EC No: 252-934-5; MDL No: MFCD00000736
UN No: UN3265; Haz Class: 8; Packing Group: II
Liquid; Linear Formula: CH3CH2OCOCH2COCl; Molecular Formula: C5H7ClO3; MW: 150.56
Boiling Point: 79-80°/25 mmHg; Flash point: 67°C (152°F)
Density (g/mL): 1.176; Refractive Index: 1.429
Moisture Sensitive
- Refractive index: 1.429
- Moisture Sensitive
- Liquid
Formula:
C₅H₇ClO₃ MW: 150.56 g/mol Boiling Pt: 79…80 °C/25 mmHg Density: 1.176 g/mL Flash Pt: 67 °C (152 °F) |
MDL Number:
MFCD00000736 CAS Number: 36239-09-5 EINECS: 252-934-5 UN: 3265 ADR: 8,II |
Specification Test Results
Assay | 95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...