Acetonedimethyl acetal ≥98%
Supplier: BeanTown Chemical
Synonyms:
DMP
222175-100ML
222175-500ML
BT222175-100MLEA
29.57
USD
BT222175-100ML
BT222175-500ML
Acetonedimethyl acetal ≥98%
Acetonedimethyl acetal
CAS: 77-76-9; EC No: 201-056-0; MDL No: MFCD00008479
UN No: UN3271; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: (CH3O)2C(CH3)2; Molecular Formula: C5H12O2; MW: 104.15
Boiling Point: 83°; Flash point: -10°C (14°F)
Density (g/mL): 0.847; Refractive Index: 1.378
- Refractive index: 1.378
- Liquid
Formula:
C₅H₁₂O₂ MW: 104.15 g/mol Boiling Pt: 83 °C Density: 0.847 g/mL Flash Pt: –10 °C (14 °F) |
MDL Number:
MFCD00008479 CAS Number: 77-76-9 EINECS: 201-056-0 UN: 3271 ADR: 3,II |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...