Zinc(II) telluride ≥99.99% (trace metals basis)

Supplier: BeanTown Chemical

222085-100G 222085-25G 222085-5G
BT222085-100GEA 857 USD
BT222085-100G BT222085-25G BT222085-5G
Zinc(II) telluride ≥99.99% (trace metals basis)
Zinc(II) telluride

CAS: 1315-11-3; EC No: 215-260-2; MDL No: MFCD00043079 UN No: UN3284; Haz Class: 6.1; Packing Group: II -140 Mesh Powder; Linear Formula: ZnTe; MW: 192.99 Melting Point: 1240° Density (g/mL): 6.34; Refractive Index: 3.56


  • Refractive index: 3.56
  • -140 Mesh Powder

Formula: TeZn
MW: 192.99 g/mol
Melting Pt: 1240 °C
Density: 6.34 g/mL
MDL Number: MFCD00043079
CAS Number: 1315-11-3
EINECS: 215-260-2
ADR: 6.1,II

Order Now

Specification Test Results

Assay 99.99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR