1,2-Dichlorobenzene ≥99%
Supplier: BeanTown Chemical
Synonyms:
o-Dichlorobenzene
222070-10KG
222070-2.5KG
222070-500G
BT222070-10KGEA
234
USD
BT222070-10KG
BT222070-2.5KG
BT222070-500G
1,2-Dichlorobenzene ≥99%
1,2-Dichlorobenzene
CAS: 95-50-1; EC No: 202-425-9; MDL No: MFCD00000535; RTECS: CZ4500000
UN No: UN1591; Haz Class: 6.1; Packing Group: III
Liquid; Molecular Formula: C6H4Cl2; MW: 147.00
Melting Point: -18°; Boiling Point: 178-180°; Flash point: 66°C (151°F)
Density (g/mL): 1.306; Refractive Index: 1.551
- RTECS: CZ4500000
- Refractive index: 1.551
- Liquid
Formula:
C₆H₄Cl₂ MW: 147 g/mol Boiling Pt: 178…180 °C Melting Pt: –18 °C Density: 1.306 g/mL Flash Pt: 66 °C (151 °F) |
MDL Number:
MFCD00000535 CAS Number: 95-50-1 EINECS: 202-425-9 UN: 1591 ADR: 6.1,III |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...