α-Bromo-4-nitrotoluene ≥97%
Supplier: BeanTown Chemical
Synonyms:
4-Nitrobenzyl bromide, 1-(Bromomethyl)-4-nitrobenzene, α-Bromo-p-nitrotoluene
222050-100G
222050-25G
222050-500G
BT222050-100GEA
128
USD
BT222050-100G
BT222050-25G
BT222050-500G
α-Bromo-4-nitrotoluene ≥97%
α-Bromo-4-nitrotoluene
CAS: 100-11-8; EC No: 202-820-6; MDL No: MFCD00007373; RTECS: XS7967000
UN No: UN3261; Haz Class: 8; Packing Group: II
Crystalline; Molecular Formula: C7H6BrNO2; MW: 216.03
Melting Point: 96-99°
- RTECS: XS7967000
- Crystalline
Formula:
C₇H₆BrNO₂ MW: 216.03 g/mol Melting Pt: 96…99 °C |
MDL Number:
MFCD00007373 CAS Number: 100-11-8 EINECS: 202-820-6 UN: 3261 ADR: 8,II |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...