(S)-(+)-Epichlorhydrin ≥98%

Supplier: BeanTown Chemical

Synonyms: (S)-(+)-Glycerol epichlorhydrin, (S)-(+)-3-Chloro-1,2-epoxypropane, (S)-(+)-1-Chloro-2,3-epoxypropane, (2S)-2-(chloromethyl)oxirane

222040-1G 222040-25G 222040-5G
BT222040-1GEA 52 USD
BT222040-1G BT222040-25G BT222040-5G
(S)-(+)-Epichlorhydrin ≥98%
(S)-(+)-Epichlorhydrin

CAS: 67843-74-7; MDL No: MFCD00077760; RTECS: RR0427100 UN No: UN2023; Haz Class: 6.1 (3); Packing Group: II Liquid; Molecular Formula: C3H5ClO; MW: 92.52 Flash point: 32°C (90°F) Density (g/mL): 1.18; Optical Rotation: [α]20/D +35°, c = 1 in methanol


  • RTECS: RR0427100
  • Optical rotation: [α]20/D +35°, c = 1 in methanol
  • Liquid

Formula: ClCH₂CH(O)CH₂
MW: 92.52 g/mol
Melting Pt: -48
Density: 1.18 g/mL
Flash Pt: 32 °C (90 °F)
MDL Number: MFCD00077760
CAS Number: 67843-74-7
UN: 2023
ADR: 6.1 II

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR