Platinum(II) chloride ≥98%
Supplier: BeanTown Chemical
CAS: 10025-65-7; EC No: 233-034-1; MDL No: MFCD00011181; RTECS: TP2275000
UN No: UN3260; Haz Class: 8; Packing Group: III
Crystalline/Powder; Linear Formula: PtCl2; MW: 265.99
Melting Point: 581° (decomposes)
Density (g/mL): 5.87
- RTECS: TP2275000
- Crystalline/Powder
Formula:
Cl₂Pt Melting Pt: 581 °C (dec.) Density: 5.87 g/mL |
MDL Number:
MFCD00011181 CAS Number: 10025-65-7 EINECS: 233-034-1 UN: 3260 ADR: 8,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...