Fullerene C60 ≥99.9%, powder

Supplier: BeanTown Chemical

221960-1G 221960-250MG 221960-5G
BT221960-1GEA 421 USD
BT221960-1G BT221960-250MG BT221960-5G
Fullerene C60 ≥99.9%, powder
Fullerene C60

CAS: 99685-96-8; MDL No: MFCD00282904; RTECS: LS9252500 UN No: UN1325; Haz Class: 4.1; Packing Group: III Powder; Molecular Formula: C; MW: 12.011


  • RTECS: LS9252500
  • Powder

Formula: C₆₀
MDL Number: MFCD00151408
CAS Number: 99685-96-8
UN: 1325
ADR: 4.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2c9ff03e3743861d242e7e023c84fcfa10a64ac&vid=53371479acfcc1403d111c470b60d1af20934c57&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR