Dibismuth triselenide ≥99.999%
Supplier: BeanTown Chemical
221935-10G
221935-50G
BT221935-10GEA
77
USD
BT221935-10G
BT221935-50G
Dibismuth triselenide ≥99.999%
Dibismuth triselenide
CAS: 12068-69-8; EC No: 235-104-7; MDL No: MFCD00014200
UN No: UN3077; Haz Class: 9; Packing Group: III
1-6mm Pieces ; Molecular Formula: Bi2Se3; MW: 654.84
Melting Point: 710°
- 1-6mm Pieces
Formula:
Bi₂Se₃ MW: 654.84 Melting Pt: 706 Density: 6.82 |
MDL Number:
MFCD00014200 CAS Number: 12068-69-8 EINECS: 235-104-7 UN: 3077 ADR: 6.1 III Merck Index: 12,01319 |
Specification Test Results
Assay | 99.999% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...