2-Chlorobenzonitrile ≥98%

Supplier: BeanTown Chemical

221920-1KG 221920-250G 221920-50G
BT221920-1KGEA 162 USD
BT221920-1KG BT221920-250G BT221920-50G
2-Chlorobenzonitrile ≥98%
2-Chlorobenzonitrile

CAS: 873-32-5; EC No: 212-836-5; MDL No: MFCD00001779; RTECS: DI2625000 UN No: UN3439; Haz Class: 6.1; Packing Group: III Crystalline; Molecular Formula: C7H4ClN; MW: 137.57 Melting Point: 43-46°; Boiling Point: 232°; Flash point: 122°C (252°F)


  • RTECS: DI2625000
  • Crystalline

Formula: C₇H₄ClN
Boiling Pt: 232 °C
Melting Pt: 43…46 °C
Flash Pt: 122 °C (252 °F)
MDL Number: MFCD00001779
CAS Number: 873-32-5
EINECS: 212-836-5
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR