Morpholine ≥99.0% ACS
Supplier: BeanTown Chemical
221880-4X500G
221880-500G
BT221880-4X500EA
278
USD
BT221880-4X500
BT221880-500G
Morpholine ≥99.0% ACS
Morpholine
CAS: 110-91-8; EC No: 203-815-1; MDL No: MFCD00005972; RTECS: QD6475000
UN No: UN2054; Haz Class: 8 (3); Packing Group: I
Liquid; Molecular Formula: C4H9NO; MW: 87.12
Melting Point: -7 - -5°; Boiling Point: 129°; Flash point: 31°C (88°F)
Density (g/mL): 0.996; Refractive Index: 1.454
Hygroscopic
- RTECS: QD6475000
- Refractive index: 1.454
- Hygroscopic
- Liquid
Formula:
C₄H₉NO MW: 87.12 g/mol Boiling Pt: 129 °C Melting Pt: –7 – –5 °C Density: 0.996 g/mL Flash Pt: 31 °C (88 °F) |
MDL Number:
MFCD00005972 CAS Number: 110-91-8 EINECS: 203-815-1 UN: 2054 ADR: 8,I |
Specification Test Results
Assay | 99.0% min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...