(±)-2,3-Dibromo-1-propanol ≥98%
Supplier: BeanTown Chemical
CAS: 96-13-9; EC No: 202-480-9; MDL No: MFCD00004699; RTECS: UB0175000
UN No: UN2810; Haz Class: 6.1; Packing Group: II
Liquid; Linear Formula: BrCH2CH(Br)CH2OH; Molecular Formula: C3H6Br2O; MW: 217.89
Boiling Point: 95-97°/10 mmHg
Density (g/mL): 2.12; Refractive Index: 1.559
- RTECS: UB0175000
- Refractive index: 1.559
- Liquid
Formula:
HOCH₂BrCHCH₂Br Boiling Pt: 95…97 °C/10 mmHg Density: 2.12 g/mL |
MDL Number:
MFCD00004699 CAS Number: 96-13-9 EINECS: 202-480-9 ADR: 6.1,II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...