Potassium hexachloroplatinate(IV)
Supplier: BeanTown Chemical
Synonyms:
Dipotassium hexachloroplatinate
221855-1G
221855-250MG
221855-5G
BT221855-1GEA
128
USD
BT221855-1G
BT221855-250MG
BT221855-5G
Potassium hexachloroplatinate(IV)
Potassium hexachloroplatinate(IV)
CAS: 16921-30-5; EC No: 240-979-3; MDL No: MFCD00011389; RTECS: TP1650000
UN No: UN3288; Haz Class: 6.1; Packing Group: III
Crystalline/Powder; Linear Formula: K2PtCl6; MW: 485.99
Melting Point: 250° (decomposes)
Density (g/mL): 3.5
- RTECS: TP1650000
- Crystalline/Powder
Formula:
K₂PtCl₆ MW: 485.99 g/mol Melting Pt: 250 °C (dec.) Density: 3.5 g/mL |
MDL Number:
MFCD00011389 CAS Number: 16921-30-5 EINECS: 240-979-3 UN: 3288 ADR: 6.1,III |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...